Learn More
4-Fluoroindole, 97%, Thermo Scientific Chemicals
Product Code. 15462428
Click to view available options
Quantity:
250 mg
1 g
5 g
CAS: 387-43-9 | C8H6FN | 135.141 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 387-43-9
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD00055992
InChI Key ZWKIJOPJWWZLDI-UHFFFAOYSA-N
Synonym 4-fluoroindole, 1h-indole, 4-fluoro, 4-fluoro-indole, pubchem7297, acmc-1cnb8, ksc224m5j, bidd:gt0103
PubChem CID 2774502
IUPAC Name 4-fluoro-1H-indole
SMILES C1=CC2=C(C=CN2)C(=C1)F
Specifications
Melting Point 30°C to 32°C
Boiling Point 90°C (0.4 mmHg)
Quantity 1 g
Formula Weight 135.14
Percent Purity 97%
Chemical Name or Material 4-Fluoroindole