Learn More
4-Cyanoimidazole-5-carboxamide hydrate, 97%, Thermo Scientific™
655.00 SEK - 2120.60 SEK
Chemical Identifiers
CAS 5372-23-6
Molecular Formula C5H4N4O
Molecular Weight (g/mol) 136.11
MDL Number MFCD00129664
InChI Key QBKYWNHLILCGEI-UHFFFAOYSA-N
Synonym 5-cyano-1h-imidazole-4-carboxamide, 4-cyano-5-imidazolecarboxamide, 5-cyano-3h-imidazole-4-carboxamide, 1h-imidazole-4-carboxamide, 5-cyano, 4-cyanoimidazole-5-carboxamide, acmc-20amkb, 5-cyanoimidazole-4-carboxamide, 1h-imidazole-5-carboxamide,4-cyano, 5-cyano-1h-imidazole-4-carboxamidehydrate, 1h-imidazole-4-carboxamide, 5-cyano-9ci
PubChem CID 270862
IUPAC Name 4-cyano-1H-imidazole-5-carboxamide
SMILES NC(=O)C1=C(N=CN1)C#N
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11430747
SDS View Documents
Thermo Scientific Alfa Aesar
B22535.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11440747
SDS View Documents
Thermo Scientific Alfa Aesar
B22535.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Chemical Identifiers
5372-23-6
136.11
QBKYWNHLILCGEI-UHFFFAOYSA-N
270862
NC(=O)C1=C(N=CN1)C#N
C5H4N4O
MFCD00129664
5-cyano-1h-imidazole-4-carboxamide, 4-cyano-5-imidazolecarboxamide, 5-cyano-3h-imidazole-4-carboxamide, 1h-imidazole-4-carboxamide, 5-cyano, 4-cyanoimidazole-5-carboxamide, acmc-20amkb, 5-cyanoimidazole-4-carboxamide, 1h-imidazole-5-carboxamide,4-cyano, 5-cyano-1h-imidazole-4-carboxamidehydrate, 1h-imidazole-4-carboxamide, 5-cyano-9ci
4-cyano-1H-imidazole-5-carboxamide
Specifications
5372-23-6
Odorless
MFCD00129664
5-cyano-1h-imidazole-4-carboxamide, 4-cyano-5-imidazolecarboxamide, 5-cyano-3h-imidazole-4-carboxamide, 1h-imidazole-4-carboxamide, 5-cyano, 4-cyanoimidazole-5-carboxamide, acmc-20amkb, 5-cyanoimidazole-4-carboxamide, 1h-imidazole-5-carboxamide,4-cyano, 5-cyano-1h-imidazole-4-carboxamidehydrate, 1h-imidazole-4-carboxamide, 5-cyano-9ci
NC(=O)C1=C(N=CN1)C#N
136.11
136.11 (Anhydrous)
4-Cyanoimidazole-5-carboxamide hydrate
∼272°C (decomposition)
C5H4N4O
UN3439
QBKYWNHLILCGEI-UHFFFAOYSA-N
4-cyano-1H-imidazole-5-carboxamide
270862
97%
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures