Learn More
4-Chlorocarbonylbenzeneboronic anhydride, tech. 90%, Thermo Scientific™
Product Code. p-7035138
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 332154-57-1
Molecular Formula C7H6BClO3
Molecular Weight (g/mol) 184.382
MDL Number MFCD06801674
InChI Key ARKQVAQLGLIPAU-UHFFFAOYSA-N
Synonym 4-chlorocarbonylphenylboronic acid, benzoylchloride-4-boronic acid, 4-chlorocarbonyl phenyl boronic acid, boronic acid, b-4-chlorocarbonyl phenyl, 4-chlorocarbonylphenyl boronic anhydride, p-chlorocarbonylphenylboronic acid, 4-chlorocarbonylphenylboronic boronic acid, 4-carbonochloridoyl phenylboronic acid, pubchem9545
PubChem CID 15345913
IUPAC Name (4-carbonochloridoylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)Cl)(O)O
Specifications
Melting Point >300°C
Quantity 1 g
Formula Weight 166.37
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Chlorocarbonylbenzeneboronic anhydride