Learn More
4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile, 97%, Thermo Scientific Chemicals
Product Code. 15428335
Click to view available options
Quantity:
250 mg
1 g
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 287922-71-8
Molecular Formula C5H4BrN3
Molecular Weight (g/mol) 186.01
MDL Number MFCD00103545
InChI Key NTTLCOOFUGHMJS-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole-3-carbonitrile, 1h-pyrazole-3-carbonitrile, 4-bromo-1-methyl, 4-bromo-1-methyl-pyrazole-3-carbonitrile
PubChem CID 45594322
IUPAC Name 4-bromo-1-methylpyrazole-3-carbonitrile
SMILES CN1C=C(Br)C(=N1)C#N
Specifications
Melting Point 103°C to 105°C
Quantity 250 mg
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 186.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile