Learn More
Memmert™ 4-20 mA Current Loop Interface for UF55
Temperature Output for external temperature monitoring.
Brand:  Memmert™ UF55V6
4970.66 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17579525
  • 5190.00 SEK / Each
Only null left
Estimated Shipment: 26-08-2025
Description
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
Specifications
UF55 Oven
Current Loop Interface
4 to 20 mA