missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluoroacetophenone, 96%, Thermo Scientific Chemicals

Product Code. p-7054474
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161957-61-5
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.04
MDL Number MFCD09264507
InChI Key GSLFYQYBXIVNLS-UHFFFAOYSA-N
Synonym 3'-bromo-2'-fluoroacetophenone, 1-3-bromo-2-fluorophenyl ethanone, 1-3-bromo-2-fluorophenyl ethan-1-one, 1-3-bromo-2-fluoro-phenyl-ethanone, 3-bromo-2-fluoroacetophenone, 1-3-bromo-2-fluoro-phenyl ethanone, 2'-fluoro-3'-bromo acetophenone, ethanone, 1-3-bromo-2-fluorophenyl, 1-acetyl-3-bromo-2-fluorobenzene, pubchem4095
PubChem CID 14937359
IUPAC Name 1-(3-bromo-2-fluorophenyl)ethanone
SMILES CC(=O)C1=C(F)C(Br)=CC=C1

Specifications

Quantity 250 mg
Formula Weight 217.04
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Bromo-2-fluoroacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.