Learn More
3,4-diethoxycyclobut-3-ene-1,2-dione, Thermo Scientific™
Product Code. 10497554
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Amber Glass Bottle
Unit Size:
100g
10g
25g
Chemical Identifiers
CAS 5231-87-8
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
InChI Key DFSFLZCLKYZYRD-UHFFFAOYSA-N
Synonym 3,4-diethoxy-3-cyclobutene-1,2-dione, diethyl squarate, squaric acid diethyl ester, 3-cyclobutene-1,2-dione, 3,4-diethoxy, diethoxycyclobutenedione, diethoxycyclobut-3-ene-1,2-dione, 1,2-diethoxycyclobutenedione, 3,4-diethoxy-cyclobut-3-ene-1,2-dione, pubchem9741, acmc-209kyn
PubChem CID 123228
IUPAC Name 3,4-diethoxycyclobut-3-ene-1,2-dione
SMILES CCOC1=C(C(=O)C1=O)OCC
Specifications
Density 1.1500g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 100 g
Formula Weight 170.16
Percent Purity 97%
Chemical Name or Material 3, 4-Diethoxycyclobut-3-ene-1, 2-dione