Learn More
2-Cyanobenzeneboronic acid, 96%
Product Code. p-7034908
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
CAS: 138642-62-3 | C7H6BNO2 | 146.94 g/mol
2-Cyanobenzeneboronic acid is useful in the synthesis of substitutes indenones or indanones. It is used in a rhodium-catalyzed [3+2] annulation with alkynes leading to substituted indenones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Cyanobenzeneboronic acid is useful in the synthesis of substitutes indenones or indanones. It is used in a rhodium-catalyzed [3+2] annulation with alkynes leading to substituted indenones.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 138642-62-3
Molecular Formula C7H6BNO2
Molecular Weight (g/mol) 146.94
MDL Number MFCD01632208
InChI Key NPLZNDDFVCGRAG-UHFFFAOYSA-N
Synonym 2-cyanophenyl boronic acid, 2-cyanobenzeneboronic acid, 2-cyanophenylboric acid, 2-cyanophenylboronicacid, 2-dihydroxyboranyl benzonitrile, boronic acid, b-2-cyanophenyl, 2-boronobenzonitrile, 2-cyanobenzene boronic acid, 2-cyano-phenyl-boronic acid, boronic acid, cyanophenyl
PubChem CID 2734610
IUPAC Name (2-cyanophenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1C#N
Specifications
Melting Point 250°C to 254°C
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 146.94
Percent Purity 96%
Chemical Name or Material 2-Cyanobenzeneboronic acid