Learn More
Molecular Probes™ 2',7'-Difluorofluorescein (Oregon Green™ 488)
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Oregon Green™ 488 can be used as a reference standard for Oregon Green™ 488 conjugates or a pH sensor of moderately acidic solutions (pKa ∼4.7)
Specifications
Specifications
Quantity | 10 mg |
Product Type | 2',7'-Difluorofluorescein |
Content And Storage | Store at room temperature and protect from light. |
Shipping Condition | Room Temperature |
Product Line | Oregon Green™ |