missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate, 99%, Honeywell™

Product Code. 15618430 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Glass bottle
Stainless steel can
This item is not returnable. View return policy
This item is not returnable. View return policy

Mixture of isomers, 99%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25265-77-4
Molecular Formula C12H24O3
Molecular Weight (g/mol) 216.321
InChI Key DAFHKNAQFPVRKR-UHFFFAOYSA-N
Synonym texanol, 2,2,4-trimethyl-1,3-pentanediol monoisobutyrate, chissocizer cs 12, propanoic acid, 2-methyl-, 3-hydroxy-2,2,4-trimethylpentyl ester, 2,2,4-trimethylpentane-1,3-diol monoisobutyrate, 3-hydroxy-2,2,4-trimethylpentyl isobutyrate, ccris 8966, isobutyraldehyde tishchenko trimer, propanoic acid, 2-methyl-, monoester with 2,2,4-trimethyl-1,3-pentanediol, 2,2,4-trimethyl-1,3-pentanediolmono 2-methylpropanoate
PubChem CID 6490
IUPAC Name (3-hydroxy-2,2,4-trimethylpentyl) 2-methylpropanoate
SMILES CC(C)C(C(C)(C)COC(=O)C(C)C)O

Specifications

Melting Point -50°C
Color Colorless
Boiling Point 255°C
Ignition Point 388°C
Packaging Glass bottle
Linear Formula (CH3)2CHCH[O(R)]C(CH3)2CH2O(R), R=-COCH(CH3)2 or H
Quantity 4 L
UN Number NONH for all modes of transport
Formula Weight 216.32g/mol
Percent Purity 99%
Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.