Learn More
SPEX Certiprep 1ML n-2,4-Dimethylphenyl-N
Brand:  SPEX Certiprep LCS-6279
1ML n-2,4-Dimethylphenyl-N
Product Code. 16339967
  • 340.00€ / 1mL
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Explore more special offers
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • N-2,4-Dimethylphenyl-N'-methylformamidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetonitrile