Learn More
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
Product Code. p-4812615 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
CAS: 879487-10-2 | C12H21BN2O2 | 236.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 879487-10-2
Molecular Formula C12H21BN2O2
Molecular Weight (g/mol) 236.12
MDL Number MFCD11867881
InChI Key OGYYMVGDKVJYSU-UHFFFAOYSA-N
Synonym 1-isopropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isopropyl-1h-pyrazole-4-boronic acid pinacol ester, 1-1-methylethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isopropyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-isopropyl-1h-pyrazole-4-boronic acid pinacolester, 1-isopropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-propan-2-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 4,4,5,5-tetramethyl-2-1-methylethyl pyrazol-4-yl-1,3,2-dioxaborolane, 1-isopropylpyrazole-4-boronic acid pinacol ester, 1-isopropyl-1h-pyrazole-4-boronic acid,pinacol ester
PubChem CID 46738021
IUPAC Name 1-propan-2-yl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES CC(C)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
Specifications
Melting Point 41°C to 46°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 236.12
Percent Purity 97%
Chemical Name or Material 1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.